missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ KnockOut™ DMEM/F-12
KnockOut™ DMEM/F-12
Brand: Gibco™ 12660012
33.48 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.48960kg
Description
Low osmolality medium without L-glutamine or HEPES buffer optimized for the growth of human embryonic and induced pluripotent stem cells.Specifications
Store at 2° to 8°C and protect from light. | |
Liquid | |
Phenol Red | |
Sustainable packaging | |
Gibco™ | |
Room Temperature |
Stem Cells (Embryonic), Stem Cells (iPS - Induced Pluripotent Stem) | |
KnockOut DMEM/F-12 | |
No Glutamine, No HEPES | |
cGMP for medical devices, 21 CFR Part 820 and ISO 13485 | |
500 mL |
For Research Use Only. Not for use in diagnostic procedures.