missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ KnockOut™ Serum Replacement - Multi-Species
GREENER_CHOICE

Product Code. p-6960066
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 15611882

Brand: Gibco™ A3181501

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A more defined, FBS-free medium supplement that supports the growth of pluripotent stem cells (PSCs) cultured on fibroblast feeder cells

Gibco™ KnockOut™ Serum Replacement – Multi-Species (KnockOut SR – Multi-Species) is a more defined, FBS-free medium supplement that supports the growth of pluripotent stem cells (PSCs) cultured on fibroblast feeder cells. Designed to replace fetal bovine serum (FBS) in existing protocols, this Research Use Only product uses the same KnockOut Serum Replacement formulation that has been trusted by many for almost 20 years. For pre-clinical or clinical applications, please see Cat. No. 10828010.

With KnockOut Serum Replacement – Multi-Species, you can:
  • Culture mouse, human, and other mammalian feeder-dependent PSCs with confidence
  • Rely on the improved performance of a qualified, FBS-free formulation
  • Enjoy the peace of mind of having a more stable price and supply compared to FBS

Culture mammalian PSCs with confidence
The KnockOut SR – Multi-Species formulation has been cited in more than 2000 publications. It has been shown to support feeder-dependent culture of embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) from mouse, human, and other mammalian species. For mouse and human PSCs, it has also been shown to support reprogramming, embryoid body (EB) formation, in vitro differentiation, cryopreservation, ESC derivation, and many other applications.

Rely on improved performance
KnockOut SR – Multi-Species has a more defined formulation compared to FBS, which contains many undefined and variable components. Unlike most FBS products, each lot is tested for its ability to support PSCs. Because of this, culturing mouse and human PSCs with KnockOut SR – Multi-Species leads to less differentiation and more compact colony morphology compared to FBS.

Enjoy the peace of mind of having stable price and supply
KnockOut SR – Multi-Species is not affected by the limited supply and fluctuating prices often seen with FBS. This makes it easier to plan your budget and buy what you need, especially since it is more affordable than most FBS products.

For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EU-Approved
Classification Serum-Free
Content And Storage Store in freezer (-5 to -20°C) and protect from light.
Product Type Serum Replacement
Quantity 100 mL
Sterility Sterile
Manufacturing Quality cGMP for medical devices, 21 CFR Part 820 and ISO 13485
Green Features Sustainable packaging
Lyophilized Yes
Product Line KnockOut
Shipping Condition Dry Ice
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.