Learn More
Cole-Parmer™ Koehler™ Penetrometer Precision Cone
Use these cones penetrometers. Koehler™ Penetrometer Precision Cones feature brass construction with stainless steel tips.
Brand: Cole-Parmer™ 59860-90
This item is not returnable.
View return policy
Description
- For use with Koehler Penetrometers
- 102.5g
- Conforms to ATSM D217 AND D937 NFT 60132
Specifications
Penetrometer Precision Cone | |
ATSM D217 AND D937 NFT 60132 | |
Brass/Stainless Steel |