missing translation for 'onlineSavingsMsg'
Learn More

Kojic acid, 99%, Thermo Scientific Chemicals

Product Code. 10105030
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-30-4
Molecular Formula C6H6O4
Molecular Weight (g/mol) 142.11
MDL Number MFCD00006580
InChI Key BEJNERDRQOWKJM-UHFFFAOYSA-N
Synonym kojic acid, 5-hydroxy-2-hydroxymethyl-4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl-4-pyrone, 4h-pyran-4-one, 5-hydroxy-2-hydroxymethyl, 5-hydroxy-2-hydroxymethyl pyran-4-one, acido kojico, 2-hydroxymethyl-5-hydroxy-gamma-pyrone, 2-hydroxymethyl-5-hydroxy-4h-pyran-4-one, unii-6k23f1tt52
PubChem CID 3840
ChEBI CHEBI:43572
IUPAC Name 5-hydroxy-2-(hydroxymethyl)pyran-4-one
SMILES OCC1=CC(=O)C(O)=CO1

Specifications

Melting Point 151.0°C to 156.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 18,II,57
Merck Index 15,5365
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in acetone and ethanol;,sparingly soluble in diethyl ether,ethyl acetate,,chloroform and pyridine;,insoluble in benezne
Formula Weight 142.11
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Kojic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'