missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Flasks with standard taper Necks

Single neck

Brand:  DWK Life Sciences 601000-0724

Additional Details : Weight : 0.40000kg

 View more versions of this product

Product Code. 17882811

  • £47.15 / Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket

Description

Description

  • Round bottom

Specifications

Specifications

5.1 in.
Borosilicate Glass
Round
1000 mL
24/40
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers