missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kontes™ Membrane Filtration Glassware from Restek
Used for routine filtration of corrosive liquids and removal of particles from HPLC solvents
Brand: Restek KT9538350000
This item is not returnable.
View return policy
Description
- Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
- Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
Specifications
Funnel 500mL, Flask 2000mL | |
Microfiltration Apparatus | |
Glass | |
500 mL, 2000 mL |