Learn More
DWK Life Sciences Kimble™ Borosilicate Micro Disposable Pipettes with Holder
Measure micro amounts of liquid in the laboratory using Kimble™ Borosilicate Micro Disposable Pipettes with Holder.
£57.65 - £162.24
Specifications
Certifications/Compliance USP type I, ASTM E438
Material Borosilicate Glass
Includes 1 Bulb assembly and 1 dispenser vial
Type Micropipet
Disposable Single-use
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
13015813
View Documents
DWK Life Sciences
764500-0010
41 mm
£57.65
Pack of 100
Estimated Shipment: 26-06-2025
to see stock.
Add to basket
13085803
View Documents Promotion Details
DWK Life Sciences
764500-0002
32 mm
£162.24
Pack of 100
Estimated Shipment: 16-04-2025
to see stock.
Add to basket
Description
Description
  • Disposable
  • Borosilicate glass
  • With holder
Specifications
Specifications
USP type I, ASTM E438
1 Bulb assembly and 1 dispenser vial
Single-use
Borosilicate Glass
Micropipet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences Kimble™ Borosilicate Micro Disposable Pipettes with Holder