Learn More
DWK Life Sciences Kimble™ Kontes™ Polyethylene Stoppers
Click to view available options
Standard Taper Joint:
14/10, 14/20, 14/35
19/22 ,19/38
29/26, 29/42
Unit Size:
Pack of 100
Description
Low-cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers.
- Leak-proof, air-tight seals
- Wide top flange for ease of use
Specifications
Specifications
| Standard Taper Joint | 29/26, 29/42 |
| Material | Polyethylene |
| Product Type | Plug Stopper |