missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Reaction-Jacketed Beakers
Click to view available options
Capacity (Metric):
100 mL
500 mL
Description
The open top provides easy access to concentrated residual samples.
- A conical lower interior surface eliminates spin-out when used with an egg-shaped PTFE magnetic stir bar
- Jacket has serrated 3/8 in. hose connections
- Design meets ASTM Specification E960, Type I requirements
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Capacity (English) | 3.38 oz. |
Description | Connection: 0.37 in. Serrated Hose |
Diameter (English) Inner | 1.85 in. |
Diameter (English) Outer | 3.14 in. |
Diameter (Metric) Inner | 4.7 cm |
Diameter (Metric) Outer | 8 cm |
Capacity (Metric) | 100 mL |
Material | Borosilicate Glass |
For Use With (Application) | Laboratory Applications Requiring a Variety of Temperature-Controlled Reactions and Useful in Biological and Chemical Reactions Including Titrations, Enzyme Digestions and Synthetic Organic Reactions |
Height (English) Inner | 2.87 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'