missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Serum Vials KG-33 Borosilicate Glass with Aluminum Seal
Autoclavable Kimble™ serum vials are ideally suited for the handling, containment, and storage of a variety of liquids.
£690.00
Specifications
Classification | A |
---|---|
Color | Clear |
Material | Borosilicate Glass |
Material (Seal) | Aluminum |
Certifications/Compliance | ASTM E438, Type I, Class A |
Description
Autoclavable Kimble™ serum vials are ideally suited for the handling, containment, and storage of a variety of liquids.
- Special design provides extra strength for freeze-drying applications
- Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
- Special blowback design provides ease of use for lyophillization stoppers
- Autoclavable
Aluminum seals, stoppers and septa are available separately - Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirement
Specifications
A | |
Borosilicate Glass | |
ASTM E438, Type I, Class A |
Clear | |
Aluminum |