Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column PEEK Nut for 3/16 in. Tubing
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
Brand:  DWK Life Sciences 420821-3519
Code : 17
Additional Details : Weight : 1.00000kg
Product Code. 12943521
  • £14.20 / Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
Description
Description
  • PEEK Nut for 3/16 in. OD Tubing
  • 5/16 in.-24 Thread
  • Use with the 420822-Series Flangeless Ferrules
  • Case of 1
  • Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
Specifications
For 0.18 in. O.D. Tubing
0.312 to 24 UNF
Each
PEEK
Tubing Nut