missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRTHA3B (Human) Recombinant Protein
Human KRTHA3B full-length ORF ( AAH09971, 1 a.a. - 404 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003884-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MPYNFCLPSLSCRTSCSSRPCVPPSCHGYTLPGACNIPANVSNCNWFCEGSFNGSEKETMQFLNDRLASYLEKVRQLERDNAELENLIRERSQQQEPLLCPSYQSYFKTIEELQQKILCSKSENARLVVQIDNAKLAADDFRTKYQTEQSLRQLVESDINSLRRILDELTLCRSDLEAQMESLKEELLSLKQNHEQEVNTLRCQLGDRLNVEVDAAPAVDLNQVLNETRNQYEALVETNRREVEQWFATQTEELNKQVVSSSEQLQSYQAEIIELRRTVNALEIELQAQHNLRYSLENTLTESEARYSSQLSQVQSLITNVESQLAEIRSDLERQNQEYQVLLDVRARLECEINTYRSLLESEDCKLPSNPCATTNACEKPIGSCVTNPCGPRSRCGPCNTFGY
Specifications
AAH09971 | |
keratin 33B | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KRT33B | |
GST |
3884 | |
Wheat germ expression system | |
25 μg | |
HA3II, Ha-3II, KRTHA3A, KRTHA3B, hHa3-II | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction