missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KT3 Epitope Tag Antibody, Agarose, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Bio-Techne NB600-420
Additional Details : Weight : 0.00970kg
Description
KT3 Epitope Tag Polyclonal antibody specifically detects KT3 Epitope Tag in Epitope Tag samples. It is validated for Immunoprecipitation.Specifications
KT3 Epitope Tag | |
Polyclonal | |
Immunoprecipitation 1:10-1:500 | |
KPPTPPPEPET Epitope Tag, KPPTPPPEPET Tag, KT3 Epitope Tag | |
Goats were immunized with KT3 (KPPTPPPEPET) conjugated to KLH. | |
0.1 mg | |
Primary | |
IgG |
Immunoprecipitation | |
Agarose | |
PBS with 0.09% Sodium Azide | |
Goat | |
Affinity Purified | |
RUO | |
Store at 4C. Do not freeze. | |
Epitope Tag |
For Research Use Only