Learn More
Saint Gobain Life Sciences Kynar PVDF Bag
Offer an economical alternative for analysis of noncorrosive gases. Kynar PVDF Bag features a corrosive-resistant Kynar construction for all your sampling application needs.
Brand:  Saint Gobain Life Sciences 01410-12
Product Code. 13345211
  • £69.10 / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Flexible
  • Will not contaminate sample
  • For use in gas sampling trains
  • Use for gas or liquid sample containment
  • Durable, heat-sealed seams ensure leak-proof operation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kynar Bag
PVDF
0.003 in.
12 in.
12 in.
8 L
0.070 mm
305 x 305 mm
305 mm
305 mm