missing translation for 'onlineSavingsMsg'
Learn More

Kynurenic acid, Tocris Bioscience™

Product Code. 30001063
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Broad spectrum glutamatergic antagonist

Broad spectrum EAA antagonist. Putative GPR35 ligand.

Sodium Salt also available.

Specifications

CAS 492-27-3
Molecular Formula C10H7NO3
Quantity 1 g
Synonym kynurenic acid, 4-hydroxyquinoline-2-carboxylic acid, kynurenate, quinurenic acid, transtorine, kinurenic acid, kynuronic acid, 4-oxo-1,4-dihydroquinoline-2-carboxylic acid, 4-hydroxyquinaldic acid, 4-hydroxy-2-quinolincarboxylic acid
InChI Key HCZHHEIFKROPDY-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=O)C=C(N2)C(=O)O
IUPAC Name 4-oxo-1H-quinoline-2-carboxylic acid
Molecular Weight (g/mol) 189.17
PubChem CID 3845
ChEBI CHEBI:18344
Formula Weight 189.17
Percent Purity >98%
Chemical Name or Material Kynurenic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'