missing translation for 'onlineSavingsMsg'
Learn More

L-2,4-Diaminobutyric acid dihydrochloride, 98+%, may cont. up to ca 10% monohydrochloride, Thermo Scientific Chemicals

£50.65 - £176.88

Chemical Identifiers

CAS 1883-09-6
Molecular Formula C4H12Cl2N2O2
Molecular Weight (g/mol) 191.052
MDL Number MFCD00064561
InChI Key CKAAWCHIBBNLOJ-QTNFYWBSSA-N
Synonym l-2,4-diaminobutyric acid dihydrochloride, s-2,4-diaminobutanoic acid dihydrochloride, s-+-2,4-diaminobutyric acid dihydrochloride, h-dab-oh.2hcl, 2,4-diaminobutyric acid dihydrochloride, 2s-2,4-diaminobutanoic acid dihydrochloride, s-+-2,4-diamino-n-butyric acid dihydrochloride, butanoic acid, 2,4-diamino-, dihydrochloride, 2s, d-dab.2hcl, 2s-2,4-diaminobutanoic acid, chloride, chloride
PubChem CID 2724265
IUPAC Name (2S)-2,4-diaminobutanoic acid;dihydrochloride
SMILES C(CN)C(C(=O)O)N.Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11322437
SDS View Documents
Thermo Scientific Alfa Aesar
L10093.03
1 g
£50.65
1g
Please to purchase this item. Need a web account? Register with us today!
11332437
SDS View Documents
Thermo Scientific Alfa Aesar
L10093.06
5 g
£176.88
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

missing translation for 'einecsNumber' : 217-542-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-2,4-Diaminobutyric acid dihydrochloride, 98+%, may cont. up to ca 10% monohydrochloride, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.