missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine, 99%, Thermo Scientific Chemicals
£29.45 - £83.92
Chemical Identifiers
CAS | 56-41-7 |
---|---|
Molecular Formula | C3H7NO2 |
Molecular Weight (g/mol) | 89.09 |
MDL Number | MFCD00064410 |
InChI Key | QNAYBMKLOCPYGJ-REOHCLBHSA-N |
Synonym | l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid |
PubChem CID | 5950 |
ChEBI | CHEBI:16977 |
IUPAC Name | (2S)-2-aminopropanoic acid |
SMILES | C[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10244700
|
Thermo Scientific Acros
102830250 |
25 g | Plastic Bottle |
£29.45
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10254700
|
Thermo Scientific Acros
102831000 |
100 g | Plastic Bottle |
£43.72
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10329273
|
Thermo Scientific Acros
102835000 |
500 g | Plastic Bottle |
£83.92
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-41-7 | |
89.09 | |
QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
5950 | |
(2S)-2-aminopropanoic acid |
C3H7NO2 | |
MFCD00064410 | |
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
CHEBI:16977 | |
C[C@H](N)C(O)=O |
Specifications
314.0°C to 316.0°C | |
56-41-7 | |
White | |
10ppm max. | |
10ppm max. | |
0.2% max. (105°C, 3 hrs) | |
C3H7NO2 | |
MFCD00064410 | |
+ 14.50 | |
15, 197 | |
0.1% max. | |
Solubility in water: 166.5g/l in water (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents | |
C[C@H](N)C(O)=O | |
89.09 | |
CHEBI:16977 | |
99% | |
L-Alanine |
200ppm max. | |
5.5 to 7 (2.5% soln. at 25°C) | |
200ppm max. | |
Authentic | |
99% | |
0.5% max. (TLC) | |
CH3CH(NH2)CO2H | |
04, 381 | |
300ppm max. | |
95% min. (c=10, H2O, 430nm) 1cm cell | |
l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid | |
QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
(2S)-2-aminopropanoic acid | |
5950 | |
89.09 | |
Crystalline Powder or Crystals |
Safety and Handling
Product Identifier
- L-Alanine
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title