missing translation for 'onlineSavingsMsg'
Learn More
L-Alanine, 99%, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals Product Code.: 15289305

L-Alanine, 99%, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals

Product Code. 15289305
2.5 kg
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
This item is not returnable. View return policy

Product Code. 15289305

Brand: Thermo Scientific Acros 451850025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
Synonym l-alanine, alanine, l-alpha-alanine, s-alanine, s-2-aminopropanoic acid, l-2-aminopropionic acid, 2s-2-aminopropanoic acid, h-ala-oh, l-+-alanine, l-2-aminopropanoic acid
PubChem CID 5950
ChEBI CHEBI:16977
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

Melting Point 314.0°C to 316.0°C
Assay Percent Range 99%
Linear Formula CH3CH(NH2)CO2H
Specific Rotation + 14.50
Solubility Information Solubility in water: 166.5g/l in water (25°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents
Quantity 2.5 kg
Formula Weight 89.09
Percent Purity 99%
Chemical Name or Material L-Alanine, specified according to the requirements of Ph.Eur.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0455579 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number451850025,451852500,451850010
Lot NumberA0449197 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number451850025,451852500,451850010
Lot NumberA0396988 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number451850025,451852500,451850010
Lot NumberA0403746 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number451850025,451852500,451850010
Lot NumberA0407591 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number451850025,451852500,451850010
Product Identifier
  • L-Alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Alanine, 99%, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.