missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Amethopterin hydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• L(+)-Amethopterin hydrate, is a potent inhibitor of dihydrofolate reductase
• It has tumoricidal activity
Applications
• L(+)-Amethopterin hydrate is an inhibitor of dihydrofolate reductase
• It can be used in anti-tumor studies
• It is a cell cycle arrest agent, which can inhibit synthesis of DNA, RNA, and proteins
• It can also inhibit production of pro-inflammatory cytokines
Specifications
Specifications
Melting Point | 195°C |
Color | Yellow to Orange |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 500 mg |
Merck Index | 14, 5985 |
Specific Rotation Condition | + 21.50 (20.00°C c=1,14g/l Na2CO3) |
Specific Rotation | + 21.5 |
Solubility Information | Solubility in water: insoluble in water. Other solubilities: insoluble in alcohol,chloroform,ether,slightly soluble in dilute hydrochloric acid,soluble in dilute solutions of alkali hydroxides a |
Show More |
Safety and Handling
Product Identifier
- L(+)-Amethopterin hydrate
- Danger
- Acute toxicity Category 3
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1A
- H360D-May damage the unborn child.
- H340-May cause genetic defects.
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'