missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ L-Amino Acid Oxidase (C. adamanteus)
Used for Enzyme
Brand: Abnova™ P5236.2mg
Additional Details : Weight : 0.00010kg
Description
#REF!Specifications
Enzyme Activity | |
L-Amino Acid Oxidase (C. adamanteus) | |
2 mg | |
RUO | |
L-Amino Acid Oxidase Assay | |
Snake | |
Native protein purified from Crotalus adamanteus Venom |
140kDa | |
Native protein purified from Crotalus adamanteus Venom | |
Store at 4°C. Do not freeze. This product is stable for at least 3 months. | |
L-AMINO | |
≥4 units per mg protein. One Unit oxidizes one micormole of L-leucine per minute at 25°C, pH 7.6. | |
None | |
Liquid |