missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Arabinose, ≥98%, MP Biomedicals™
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Color | White |
Density | 1.585g/mL at 20°C |
Assay Percent Range | ≥98% |
Quantity | 500 g |
Formula Weight | 150.1 |
Physical Form | Powder |
Chemical Name or Material | L-(+)-Arabinose |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction