missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Arabinose, ≥98%, MP Biomedicals™
£146.00 - £477.00
Chemical Identifiers
CAS | 5328-37-0 |
---|---|
Molecular Formula | C5H10O5 |
Molecular Weight (g/mol) | 150.13 |
InChI Key | PYMYPHUHKUWMLA-VAYJURFESA-N |
Synonym | l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l |
PubChem CID | 5460291 |
ChEBI | CHEBI:6182 |
IUPAC Name | (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal |
SMILES | C(C(C(C(C=O)O)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11417800
|
MP Biomedicals
0210070680 |
100 g |
£146.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427800
|
MP Biomedicals
0210070690 |
500 g |
£477.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
5328-37-0 | |
150.13 | |
l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l | |
CHEBI:6182 | |
C(C(C(C(C=O)O)O)O)O |
C5H10O5 | |
PYMYPHUHKUWMLA-VAYJURFESA-N | |
5460291 | |
(2R,3S,4S)-2,3,4,5-tetrahydroxypentanal |
Specifications
5328-37-0 | |
1.585g/mL at 20°C | |
C5H10O5 | |
PYMYPHUHKUWMLA-VAYJURFESA-N | |
(2R,3S,4S)-2,3,4,5-tetrahydroxypentanal | |
5460291 | |
150.1 | |
L-(+)-Arabinose |
White | |
≥98% | |
l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l | |
C(C(C(C(C=O)O)O)O)O | |
150.13 | |
CHEBI:6182 | |
Powder |
Safety and Handling
Recommended Storage : Store at room temperature.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title