missing translation for 'onlineSavingsMsg'
Learn More
L(+)-Arginine, 98+%, Thermo Scientific Chemicals Product Code.: 10115003

L(+)-Arginine, 98+%, Thermo Scientific Chemicals

Product Code. 10115003
2.5 kg, Plastic Bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10115003

Brand: Thermo Scientific 104990025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98+%
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Plastic Bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H
Specific Rotation +25.8° to +28.5° (20°C, 589nm) (c=8, 6N HCl) on dried substance
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Specific Rotation Condition +27.10° (20°C c=8,6N HCl)
Quantity 2.5 kg
Formula Weight 174.2
Percent Purity ≥98.0% (on dry substance)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Arginine, 98+%
Show More Show Less
compliance-icons
Identificativo prodotto
  • L(+)-Arginine
Avvertenza
  • Warning
Categoria di pericolo
  • Serious eye damage/eye irritation Category 2
Indicazioni di pericolo
  • H319-Causes serious eye irritation.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
L(+)-Arginine, 98+%, Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato