missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Arginine, 98+%

Product Code. p-60848 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
100g
10kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10664752

Brand: Thermo Scientific Acros 104991000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 74-79-3 | C6H14N4O2 | 174.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES NC(CCCN=C(N)N)C(O)=O

Specifications

Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98+%
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Plastic Bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NH2)CO2H
Specific Rotation +25.8° to +28.5° (20°C, 589nm) (c=8, 6N HCl) on dried substance
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Quantity 100 g
Formula Weight 174.2
Percent Purity ≥98.0% (on dry substance)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Arginine, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Arginine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.