missing translation for 'onlineSavingsMsg'
Learn More

L-Arginine Hydrochloride, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15537954
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Packaging Poly Bottle
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
Molecular Weight (g/mol) 210.662
PubChem CID 66250
Formula Weight 210.66
Grade Multi-Compendial/USP
Quantity 1 kg
Specific Gravity 1L = 1.42kg
Chemical Name or Material L-Arginine Hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'