missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Arginine monohydrochloride, 98+% Product Code.: 11477766

Thermo Scientific Chemicals L-Arginine monohydrochloride, 98+%

Product Code. 11477766
2500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11477766

Brand: Thermo Scientific Chemicals A14730.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Arginine monohydrochloride (L-Arginine hydrochloride or arginine HCl, CAS # 1119-34-2) is the salt of the essential amino acid arginine.

L-Arginine monohydrochloride is a precursor of nitric oxide synthesis that induces vasodilation, there by relaxing blood vessels. It is an important intermediate in the urea cycle and involved in the detoxification of nitrogenous wastes. It is commonly used in cell culture media and drug development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Arginine is the left-handed enantiomer of the amino acid arginine, an essential amino acid in juvenile humans
  • Arginine residues are common active site components in enzymes
  • Arginine is involved in nitric acid production, and many biological processes depend on it
  • Arginine plays a role in the immune system and in the detoxification of nitrogenous wastes

Application

  • L-Arginine monohydrochloride is suitable for biochemistry and in vitro research
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-34-2
Molecular Formula C6H15ClN4O2
Molecular Weight (g/mol) 210.662
MDL Number MFCD00064550
InChI Key KWTQSFXGGICVPE-WCCKRBBISA-N
Synonym l-arginine hydrochloride, arginine hydrochloride, l-arginine hcl, l-arginine monohydrochloride, h-arg-oh.hcl, r-gene, arginine monohydrochloride, argamine, l-arginine, monohydrochloride, unii-f7lth1e20y
PubChem CID 66250
IUPAC Name (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN=C(N)N.Cl

Specifications

Melting Point 216°C (decomposition)
Beilstein 3631658
Merck Index 14,780
Solubility Information Soluble in water. Slightly soluble in hot alcohol.
Density 1.42
Quantity 2500 g
Formula Weight 210.67
Optical Rotation +22° (c=5 in 5N HCl)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Arginine monohydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249981 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog NumberA14730.22,A14730.36,A14730.0E
Lot Number10247860 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog NumberA14730.22,A14730.36,A14730.0E
Lot Number10241486 Certificate TypeCertificate of Analysis Date01/07/2022 Catalog NumberA14730.22,A14730.36,A14730.0E
Lot Number10238574 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14730.22,A14730.36,A14730.0E
Lot Number10234716 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14730.22,A14730.36,A14730.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Arginine monohydrochloride, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.