missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic acid 6-palmitate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
L-Ascorbyl palmitate (Asc-6P) is used along with other fatty acid ascorbates such as L-ascorbyl oleate and L-ascorbyl linoleate to protect cells and membranes from oxidative damage. Fatty acid ascorbate molecules have increased solubility in non-polar (lipophilic) environments. It is applied as humectants and emollients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 113°C to 114°C |
Odor | Pleasant |
Quantity | 25 g |
Solubility Information | Slightly soluble in ethyl alcohol. |
Formula Weight | 414.52 |
Percent Purity | 99% |
Physical Form | Crystalline |
Chemical Name or Material | L-Ascorbic acid 6-palmitate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'