missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid, 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 11458952
Click to view available options
Quantity:
100 g
500 g
1 kg

L-Ascorbic Acid, 99+%, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
MDL Number 64328

Specifications

Melting Point 190°C
Color White
pH 2.1 to 2.6
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 176.13g/mol
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'