missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic Acid ACS AR, Macron Fine Chemicals™
Click to view available options
Quantity:
500 g
Packaging:
Glass
Specifications
Specifications
Packaging | Glass |
Quantity | 500 g |
IUPAC Name | (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one |
Formula Weight | 176.13 |
Grade | ACS |
Chemical Name or Material | L-Ascorbic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'