missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ascorbic acid, ACS reagent

Product Code. 10055800 Shop All Thermo Scientific Chemicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
missing translation for 'unitSize'
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10055800

missing translation for 'mfr': Thermo Scientific Acros 401471000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 50-81-7 | C6H8O6 | 176.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point 190.0°C to 192.0°C
Color White to Yellow
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 100 g
Beilstein 18, IV, 3038
Merck Index 15, 819
Specific Rotation + 20.50
Solubility Information Solubility in water: 333g/L (20°C). Other solubilities: ethanol 20g/L (20°C), soluble methanol and acetone, insoluble in ether, chloroform and benzene
Formula Weight 176.13
Percent Purity ≥99.0%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material L(+)-Ascorbic acid, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.