missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic Acid, Certified AR for Analysis, Fisher Chemical™
141.16 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Description
C6H8O6, CAS Number-50-81-7, 500g, White, 176.13g/mol, 190 deg.C, 64328, Plastic powder jar, 2.1 to 2.6, Solid
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 190°C |
Color | White |
pH | 2.1 to 2.6 |
Packaging | Plastic powder jar |
Quantity | 500 g |
Formula Weight | 176.13g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'