missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Ascorbic acid sodium salt, 99%

£27.24 - £390.95

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00082340
InChI Key WHPNKQBYKGWBQD-PQYRJTSOSA-N
Synonym sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
PubChem CID 131674100
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one;molecular hydrogen;sodium
SMILES [HH].C(C(C1C(=C(C(=O)O1)O)O)O)O.[Na]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10012011
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Chemicals
352680050
5 g Glass bottle
£27.24
5g
Please to purchase this item. Need a web account? Register with us today!
10113731
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
352681000
100 g Plastic bottle
£37.89
100g
Please to purchase this item. Need a web account? Register with us today!
10123731
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
352685000
500 g Plastic bottle
£123.62
500g
Please to purchase this item. Need a web account? Register with us today!
11946881
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
352680025
2.5 kg Plastic bottle
£390.95
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

134-03-2
198.11
WHPNKQBYKGWBQD-PQYRJTSOSA-N
131674100
[HH].C(C(C1C(=C(C(=O)O1)O)O)O)O.[Na]
C6H7NaO6
MFCD00082340
sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one;molecular hydrogen;sodium

Specifications

134-03-2
White to Yellow
98.5 to 101.5% (Iodimetry)
MFCD00082340
+ 104.00
(5% in water) Clear colorless to light yellow
[HH].C(C(C1C(=C(C(=O)O1)O)O)O)O.[Na]
198.11
198.11
Crystalline Powder
219.0°C to 221.0°C
Authentic
C6H7NaO6
15,819
sodium ascorbate, l-ascorbic acid sodium salt, sodium l-ascorbate, vitamin c sodium, ascorbicin, sodascorbate, cebitate, aminofenitrooxon, natrii ascorbas, monosodium l-ascorbate
WHPNKQBYKGWBQD-PQYRJTSOSA-N
(2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one;molecular hydrogen;sodium
131674100
99%
L-Ascorbic acid sodium salt, 99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-126-1

missing translation for 'rtecsNumber' : CI7671000

missing translation for 'tsca' : TSCA

Product Identifier
  • L-Ascorbic acid sodium salt

SDS
Documents

Documents

Certificates

RUO – Research Use Only