missing translation for 'onlineSavingsMsg'
Learn More

L-Ascorbic Acid (White Crystalline Powder), Fisher BioReagents

Product Code. 10061793
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10061793

Brand: Fisher BioReagents BP351500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Ascorbic Acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Ascorbic Acid
CAS 50-81-7
Melting Point 190°C
Color Yellow
Density 1.65g/cm³
Ignition Residue 0.1% max.
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Molecular Formula C6H8O6
MDL Number MFCD00064328
Quantity 500 g
Merck Index 15, 819
Specific Rotation 20.5 to 21.5° (+ or -)
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Molecular Weight (g/mol) 176.12
PubChem CID 54670067
ChEBI CHEBI:29073
Formula Weight 176.13
Percent Purity ≥99.0%
Physical Form Solid
Chemical Name or Material L-Ascorbic Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.