missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 111°C to 115°C |
Boiling Point | 250°C |
Ignition Residue | 0.1% max |
Packaging | Glass bottle |
Quantity | 100 g |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 414.53 |
Chemical Name or Material | L(+)-Ascorbyl palmitate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'