missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Asparagine, 99%, Thermo Scientific Chemicals
£72.54 - £930.63
Chemical Identifiers
CAS | 70-47-3 |
---|---|
Molecular Formula | C4H8N2O3 |
Molecular Weight (g/mol) | 132.119 |
MDL Number | MFCD00064401 |
InChI Key | DCXYFEDJOCDNAF-REOHCLBHSA-N |
Synonym | l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid |
PubChem CID | 6267 |
ChEBI | CHEBI:17196 |
IUPAC Name | (2S)-2,4-diamino-4-oxobutanoic acid |
SMILES | C(C(C(=O)O)N)C(=O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10109382
|
Thermo Scientific Acros
371601000 |
100 g | Glass Bottle |
£72.54
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10537193
|
Thermo Scientific Acros
371605000 |
500 g | Glass Bottle |
£288.44
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10564654
|
Thermo Scientific Acros
371600025 |
2.5 kg | Plastic Bottle |
£930.63
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
70-47-3 | |
132.119 | |
DCXYFEDJOCDNAF-REOHCLBHSA-N | |
6267 | |
(2S)-2,4-diamino-4-oxobutanoic acid |
C4H8N2O3 | |
MFCD00064401 | |
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid | |
CHEBI:17196 | |
C(C(C(=O)O)N)C(=O)N |
Specifications
70-47-3 | |
Authentic | |
C4H8N2O3 | |
MFCD00064401 | |
l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid | |
1% max. | |
C(C(C(=O)O)N)C(=O)N | |
+ 34.50 (20.00°C c=10,2N HCl) | |
6267 | |
132.12 | |
Crystalline Powder |
White | |
99% | |
H2NCOCH2CH(NH2)CO2H | |
+ 34.50 | |
Solubility in water: 20g/l (20°C). Other solubilities: insoluble in organic solvents | |
DCXYFEDJOCDNAF-REOHCLBHSA-N | |
(2S)-2,4-diamino-4-oxobutanoic acid | |
132.119 | |
CHEBI:17196 | |
99% | |
L-Asparagine |
Safety and Handling
Product Identifier
- L-Asparagine
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Asparagine, 99%, Thermo Scientific Chemicals