missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Asparagine anhydrous, 100%, MP Biomedicals™
Click to view available options
Quantity:
25 g
Specifications
CAS | 70-47-3 |
Color | White |
Assay Percent Range | 1 |
Molecular Formula | C4H8N2O3 |
Synonym | l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid |
InChI Key | DCXYFEDJOCDNAF-REOHCLBHSA-N |
SMILES | C(C(C(=O)O)N)C(=O)N |
IUPAC Name | (2S)-2,4-diamino-4-oxobutanoic acid |
Molecular Weight (g/mol) | 132.119 |
PubChem CID | 6267 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'