missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15439798
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17812-32-7
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00080289
InChI Key SWWBMHIMADRNIK-UHFFFAOYNA-N
Synonym h-asp-ome, 1-methyl l-aspartate, l-aspartic acid 1-methyl ester, s-3-amino-4-methoxy-4-oxobutanoic acid, 3s-3-amino-4-methoxy-4-oxobutanoic acid, l-aspartic acid methyl ester, pubchem13176, l-aspartic acid 1-methyl ester; 3-amino-4-methoxy-4-oxobutanoic acid; h-asp-ome
PubChem CID 6994682
SMILES COC(=O)C(N)CC(O)=O

Specifications

Melting Point 186°C (decomposition)
Color White
Beilstein 2080763
IUPAC Name 3-amino-4-methoxy-4-oxobutanoic acid
Quantity 1 g
Formula Weight 147.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Aspartic acid 1-methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'