missing translation for 'onlineSavingsMsg'
Learn More
L(+)-Aspartic acid, 98+%, Thermo Scientific Chemicals Product Code.: 10714981

L(+)-Aspartic acid, 98+%, Thermo Scientific Chemicals

Product Code. 10714981
100 g, Plastic Bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10714981

Brand: Thermo Scientific Acros 105041000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acidShow MoreShow Less
PubChem CID 5960
ChEBI CHEBI:17053
IUPAC Name (2S)-2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point >230.0°C
Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98+%
Loss on Drying 0.5% max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Linear Formula HO2CCH2CH(NH2)CO2H
Specific Rotation + 25.00
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 5g/L water (25°C). Other solubilities: 2.1g/L water (0°C), 8.5g/L water (40°C), 28.7g/L water (75°C), 16 mg/100 mL ethanol (25°C), soluble in acids, alkalies
Quantity 100 g
Formula Weight 133.1
Percent Purity ≥98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Aspartic acid, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468729 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number105041000,105040025,105040100,105045000
Lot NumberA0461907 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number105041000,105040025,105040100,105045000
Lot NumberA0392808 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number105041000,105040025,105040100,105045000
Lot NumberA0404516 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number105041000,105040025,105040100,105045000
Lot NumberA0409821 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number105041000,105040025,105040100,105045000
Product Identifier
  • L(+)-Aspartic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L(+)-Aspartic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.