missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Aspartic acid, 98+%

Product Code. 15169153 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
100g
10kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15169153

Brand: Thermo Scientific Acros 105040100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-84-8 | C4H7NO4 | 133.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-84-8
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.10
MDL Number MFCD00002616
InChI Key CKLJMWTZIZZHCS-UHFFFAOYNA-N
Synonym l-aspartic acid, aspartic acid, h-asp-oh, l-aspartate, asparagic acid, aspatofort, l-asparagic acid, l-aminosuccinic acid, s-2-aminosuccinic acid, asparaginic acid
PubChem CID 5960
ChEBI CHEBI:17053
IUPAC Name (2S)-2-aminobutanedioic acid
SMILES NC(CC(O)=O)C(O)=O

Specifications

Melting Point >230.0°C
Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98+%
Loss on Drying 0.5% max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Drum
Linear Formula HO2CCH2CH(NH2)CO2H
Specific Rotation + 25.00
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 5g/L water (25°C). Other solubilities: 2.1g/L water (0°C), 8.5g/L water (40°C), 28.7g/L water (75°C), 16 mg/100 mL ethanol (25°C), soluble in acids, alkalies
Specific Rotation Condition + 25.00 (20.00°C c=8,6N HCl)
Quantity 10 kg
Formula Weight 133.1
Percent Purity ≥98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Aspartic acid, 98+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.