missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Aspartic acid, 98+%
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
Description
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
Solubility
Soluble in water(4.5g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
L-Aspartic acid is used in enzyme active centers, as well as in maintaining the solubility and ionic character of proteins.
Solubility
Soluble in water(4.5g/L).
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
| Melting Point | >300°C (decomposition) |
| Beilstein | 1723530 |
| Merck Index | 14,840 |
| Solubility Information | Soluble in water(4.5g/L). |
| Density | 1.66 |
| IUPAC Name | (2S)-2-aminobutanedioic acid |
| Quantity | 1000 g |
| Odor | Odorless |
| Formula Weight | 133.1 |
| Optical Rotation | +25° (c=2 in 5N HCl) |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction