missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid diethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

£20.70 - £141.00

Chemical Identifiers

CAS 16115-68-7
Molecular Formula C8H16ClNO4
Molecular Weight (g/mol) 225.669
MDL Number MFCD00038910
InChI Key AJOXZAAREAYBQR-RGMNGODLSA-N
Synonym l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
PubChem CID 12416466
IUPAC Name diethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES CCOC(=O)CC(C(=O)OCC)N.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15470969
View Documents
Thermo Scientific Alfa Aesar
H63974.06
5 g
£20.70
5g
Please to purchase this item. Need a web account? Register with us today!
15480969
View Documents
Thermo Scientific Alfa Aesar
H63974.14
25 g
£66.40
25g
Please to purchase this item. Need a web account? Register with us today!
15418616
View Documents
Thermo Scientific Alfa Aesar
H63974.22
100 g
£141.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16115-68-7
225.669
AJOXZAAREAYBQR-RGMNGODLSA-N
12416466
CCOC(=O)CC(C(=O)OCC)N.Cl
C8H16ClNO4
MFCD00038910
l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
diethyl (2S)-2-aminobutanedioate;hydrochloride

Specifications

16115-68-7
MFCD00038910
AJOXZAAREAYBQR-RGMNGODLSA-N
diethyl (2S)-2-aminobutanedioate;hydrochloride
12416466
98%
L-Aspartic acid diethyl ester hydrochloride
C8H16ClNO4
l-aspartic acid diethyl ester hydrochloride, h-asp oet-oet.hcl, h-asp oet-oet hcl, s-diethyl 2-aminosuccinate hydrochloride, aspartic acid, diethyl ester, hydrochloride, h-asp oet-oet hydrochloride, diethyl l-aspartate hydrochloride, 1,4-diethyl 2s-2-aminobutanedioate hydrochloride, h-asp oet-oet?cl, diethylaspartate hydrochloride
CCOC(=O)CC(C(=O)OCC)N.Cl
225.669
225.67
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only