missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Carvone, 99%, Thermo Scientific Chemicals

Product Code. 10041810
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6485-40-1
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001578
InChI Key ULDHMXUKGWMISQ-SECBINFHSA-N
Synonym --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone
PubChem CID 439570
ChEBI CHEBI:15400
IUPAC Name (5R)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC(=C)[C@@H]1CC=C(C)C(=O)C1

Specifications

Color Colorless to Yellow
Density 0.9580g/mL
Boiling Point 227.0°C to 230.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.5020
Quantity 500 g
Beilstein 07, 157
Merck Index 14, 1874
Specific Gravity 0.958
Specific Rotation − 57.00
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in alcohol, ether, chloroform,, propylene glycol and mineral oils, insoluble in glycerol
Formula Weight 150.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material L(-)-Carvone, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Carvone
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.