missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Citrulline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.
Solubility
It is soluble in water.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
L-Citrulline, is used as an essential intermediate in the biosynthesis of nitric oxide from L-arginine . It is also used as a nutritional drink and biochemical reagent.
Solubility
It is soluble in water.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 212°C to 214°C (decomposition) |
Quantity | 10 g |
Beilstein | 6055157 |
Merck Index | 14,2331 |
Solubility Information | It is soluble in water. |
Optical Rotation | +22° (c=2 in 1N HCl) |
Formula Weight | 175.19 |
Percent Purity | 98% |
Chemical Name or Material | L-Citrulline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'