missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Citrulline, 99%, Thermo Scientific Chemicals

Product Code. 10004013
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-75-8
Molecular Formula C6H13N3O3
Molecular Weight (g/mol) 175.19
MDL Number MFCD00064397
InChI Key RHGKLRLOHDJJDR-UHFFFAOYNA-N
Synonym l-citrulline, citrulline, h-cit-oh, delta-ureidonorvaline, sitrulline, n5-carbamoyl-l-ornithine, l-cytrulline, n5-aminocarbonyl ornithine, n delta-carbamylornithine, citrulline, l
PubChem CID 9750
ChEBI CHEBI:16349
IUPAC Name (2S)-2-amino-5-(carbamoylamino)pentanoic acid
SMILES NC(CCCNC(N)=O)C(O)=O

Specifications

Melting Point 214°C
Color White
Ignition Residue 0.1% max.
Loss on Drying 0.3% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NCONH(CH2)3CH(NH2)CO2H
Quantity 100 g
Merck Index 15, 2330
Specific Rotation Condition + 25.50 (20.00°C c=8,6N HCl)
Specific Rotation + 25.5
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: insoluble in methanol and ethanol
Transmittance 95.0% min. (c=10, H2O, 430nm) 1cm cell
Formula Weight 175.19
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Citrulline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'