missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine Hydrochloride Monohydrate, 98.5 to 101.5% (Dry Basis), Thermo Scientific Chemicals

Catalog Number p-7082816
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
7048-04-6
C3H10ClNO3S
175.63
MFCD00065606
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
(2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
C(C(C(=O)O)N)S.O.Cl
This item is not returnable. View return policy
7048-04-6
C3H10ClNO3S
175.63
MFCD00065606
QIJRTFXNRTXDIP-JIZZDEOASA-N
l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.H2O, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
23462
(2 R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
C(C(C(=O)O)N)S.O.Cl
Melting Point ∼175°C (decomposition)
Linear Formula HSCH2CH(NH2)COOH.HCl.H2O
Beilstein 5158059
Packaging Plastic Bottle
Formula Weight 175.63
Percent Purity 98.5 to 101.5%
Quantity 25 g
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Cysteine Hydrochloride Monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'