missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Cysteine hydrochloride monohydrate, 99%

Product Code. 11468563
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11468563

Brand: Thermo Scientific Chemicals A10389.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7048-04-6 | C3H10ClNO3S | 175.63 g/mol

L-Cysteine hydrochloride monohydrate is used as a precursor in food, pharmaceutical and personal care products. It is actively involved in the production of flavors such as meat flavor obtained by reacting with sugars. It plays an important role as a key raw material for the manufacture of N-acetyl-L-cysteine, S-carboxymethyl-L-cysteine and L-cysteine base. It is involved in the treatment of liver disease as an antioxidant and antidote. Being an essential amino acid, it used as animal feed especially for sheep, which is responsible the growth of wool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in cold water.

Notes
Air and light sensitive. Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, metals and hydrogen chloride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
MDL Number MFCD00065606
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
Synonym l-cysteine hydrochloride monohydrate, h-cys-oh.hcl.h2o, l-cysteine hydrochloride hydrate, l-cysteine monohydrate monochloride, r-2-amino-3-mercaptopropanoic acid hydrochloride hydrate, unii-zt934n0x4w, l-cysteine hydrate hydrochloride, cysteine hydrochloride monohydrate, l, h-cys-ohhclh2o, cysteine hcl
PubChem CID 23462
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
SMILES C(C(C(=O)O)N)S.O.Cl

Specifications

Melting Point ∼175°C (decomposition)
pH 1 to 2
Beilstein 5158059
Merck Index 14,2781
Solubility Information Soluble in cold water.
Quantity 500 g
Formula Weight 175.64 (157.62 Anhydrous)
Optical Rotation +5.5° (c=5 in 5N HCl)
Sensitivity Air Sensitive
Percent Purity 99%
Chemical Name or Material L-Cysteine hydrochloride monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • L-Cysteine hydrochloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : L-Cysteine hydrochloride monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.