missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteine Hydrochloride, Monohydrate, Multi-Compendial, U.S.P., J.T. Baker™
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Formula Weight | 175.64 |
Grade | Multi-Compendial/USP |
Quantity | 1 kg |
Specific Gravity | 1L = 1.50kg |
Chemical Name or Material | L-Cysteine Hydrochloride, Monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'