missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cystine, Cell Culture Reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Description
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.
Solubility
It is soluble in water and HCL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.
Solubility
It is soluble in water and HCL.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
Specifications
Melting Point | >240°C (decomposition) |
Color | White |
Beilstein | 1728094 |
Merck Index | 14,2782 |
Solubility Information | It is soluble in water and HCL. |
Formula Weight | 240.3 |
Grade | Cell Culture Reagent |
Density | 1.68 |
Quantity | 10 g |
Optical Rotation | −219° (c=1 in 1N HCl) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'