missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cystine dihydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.
Solubility
Freely soluble in Dilute hydrochloric acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.
Solubility
Freely soluble in Dilute hydrochloric acid.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 228°C to 232°C |
Color | White to Yellow |
Solubility Information | Freely soluble in Dilute hydrochloric acid. |
Formula Weight | 313.22 |
Grade | Cell Culture Reagent |
Quantity | 250 g |
Physical Form | Powder |
Chemical Name or Material | L-Cystine dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'