missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Dihydroorotic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
L-Dihydroorotic acid a metabolite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Dihydroorotic acid a metabolite.
Solubility
Soluble in water (partly), and dimethyl formamide.
Notes
Store at room temperature. Keep away from oxidizing agents.
L-Dihydroorotic acid a metabolite.
Solubility
Soluble in water (partly), and dimethyl formamide.
Notes
Store at room temperature. Keep away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 254°C to 255°C (decomposition) |
Quantity | 5 g |
Solubility Information | Soluble in water (partly),and dimethyl formamide. |
Formula Weight | 158.11 |
Percent Purity | 98% |
Chemical Name or Material | L-Dihydroorotic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'